missing translation for 'onlineSavingsMsg'
Learn More

I-BET762 carboxylic acid, MedChemExpress

Product Code. p-200096260
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16440487 50 mg 50mg
16431089 1 mg 1mg
16416837 5 mg 5mg
16404138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16440487 Supplier Medchem Express Supplier No. HY107443

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress I-BET762 carboxylic acid (Molibresib carboxylic acid) is an I-BET762-based warhead ligand for conjugation reactions of PROTAC targeting on BET. I-BET762 carboxylic acid (Molibresib carboxylic acid) is a BRD4 inhibitor with a pIC50 of 5.1.

I-BET762 carboxylic acid (Molibresib carboxylic acid) is an I-BET762-based warhead ligand for conjugation reactions of PROTAC targeting on BET. I-BET762 carboxylic acid (Molibresib carboxylic acid) is a BRD4 inhibitor with a pIC50 of 5.1.

Specifications

Chemical Name or Material I-BET762 carboxylic acid
CAS 1300019-38-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17ClN4O3
Quantity 50 mg
Synonym Molibresib carboxylic acid GSK525762A carboxylic acid PROTAC BRD4-binding moiety 2
Solubility Information DMSO : 7.14 mg/mL (17.99 mM; Need ultrasonic)
SMILES CC1=NN=C2[C@H](CC(O)=O)N=C(C3=CC=C(Cl)C=C3)C4=C(C=CC(OC)=C4)N21
Molecular Weight (g/mol) 396.83
Formula Weight 396.83
Percent Purity 98.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.