missing translation for 'onlineSavingsMsg'
Learn More

hydrogen tetrachloroaurate(III) hydrate

Product Code. 10001611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001611 5 g Glass bottle 5g
10791111 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001611 Supplier Thermo Scientific Acros Supplier No. 223620050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 27988-77-8 | AuCl4H | 339.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27988-77-8
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149903
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Synonym Chloroauric acid hydrate, Gold trichloride acid hydrate
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Specifications

Melting Point 30.0°C
Color Orange
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Linear Formula HAuCl4·xH2O
Merck Index 14, 4522
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 339.78
Percent Purity ≥49% (Au)
Packaging Glass bottle
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen tetrachloroaurate(III) hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.