missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT™, J.T. Baker™

Product Code. 10107860 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
50 mL
150 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10107860 150 mL Each
10210051 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107860 Supplier JT Baker Supplier No. 4657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Laboratory, Research, or Manufacturing Use

  • Management of Change (MOC) category = N
  • (1/10 equiv. = 3.646 g HCl)
  • Makes 0.1 N solution after dilution to 1000 mL

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 0.1 mol/L
Density 1 g/cm³ (20°C)
Packaging DILUT-IT™ Ampule (LDPE)
Normality Passes Test
Quantity 150 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.