missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid 36 - 38%, BAKER ANALYZED™, J.T. Baker™
79.19 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -30°C |
Boiling Point | 110°C (1013 hPa) |
Density | 1.18 g/cm³ (20°C) |
Residue after Ignition | max. 5 ppm |
Assay Percent Range | 37 to 38% |
Quantity | 2.5 L |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'