missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4HEK293 Powdered Media

Product Code. 10175873
Click to view available options
Quantity:
5 L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 10175873

Brand: Cytiva SH30522.01

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Promotes adenovirus and recombinant protein production in HEK 293 cell culture.

  • Optimized for high cell growth and high recombinant protein and vector production with human embryonic kidney (HEK) 293 cells
  • Protein-free and animal-derived component-free medium available in liquid and powder form
  • Designed for small- and large-scale biomanufacturing processes
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Supports direct or sequential adaptation
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
Without Additives No Glutamine, No Phenol Red, No Poloxamer 188, No Sodium Bicarbonate
Quantity 5 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.