Learn More
Invitrogen™ Human TNF-alpha Recombinant Protein
Description
The mature, soluble form of human TNFa was expressed in Pichia pastoris, KM71H. GenBank Accession # P01375, amino acids 77 - 233. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: VRSSSRTPSD. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant human TNFa was measured by its cytolytic effect on mouse L-929 cells in the presence of Actinomycin D. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 1 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.
Specifications
Specifications
| Accession Number | P01375 |
| For Use With (Application) | Bioactivity |
| Formulation | PBS with no preservative |
| Gene ID (Entrez) | 7124 |
| Molecular Weight (g/mol) | 17.4 kDa |
| Name | Human TNF-alpha |
| Purification Method | Purified |
| Quantity | 20 μg |
| Source | Yeast |
| Storage Requirements | 4°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.