missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat alpha-Cardiac Actin Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18725189
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18725189 100 μg 100µg
18799667 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18725189 Supplier R&D Systems Supplier No. MAB9308100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin Alpha 1 Cardiac Muscle
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 959606
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P68032
Gene Alias ACTC, ACTC1, actin, alpha, cardiac muscle 1, ASD5, Cardiac alpha-Actin, CMD1R, CMH11, LVNC4
Host Species Mouse
Immunogen E.coli-derived recombinant human alpha-Cardiac Actin Asp3-Phe377 Accession # P68032
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70
Test Specificity Detects human alpha-Cardiac Actin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.