missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-alpha A (alpha 2a) Recombinant Protein

Product Code. 10180863
Change view
Click to view available options
Quantity:
8.4 x 10^6 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10180863 8.4 x 10^6 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10180863 Supplier Gibco™ Supplier No. PHC4814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Dilution Recommendation: Further dilutions should be made in medium or buffered solution containing carrier protein such as PBS with 0.1% BSA.

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01563
For Use With (Application) Bioactivity
Formulation PBS with 0.1% BSA and no preservative
Gene ID (Entrez) 3440
Molecular Weight (g/mol) 19.2 kDa
Name Human IFN-alpha A (alpha 2a)
Purification Method Purified
Quantity 8.4 x 10^6 units
Storage Requirements -80°C
Regulatory Status RUO
Gene Alias alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol IFNA2
Biological Activity Specific activity = 4.0 x 10e8 units/mg; Assay used to measure bioactivity: interferon was titrated with the use of the cytopathic effect inhibition assay as described [Rubinstein, S., Familletti, P.C., and Pestka, S. (1981'Convenient Assay for Interferons,' J. Virol. 37, 755-758; Familletti, P.C., Rubinstein, S., and Pestka, S. (1981)'A Convenient and Rapid Cytopathic Effect Inhibition Assay for Interferon,' in Methods in Enzymology, Vol. 78 (S. Pestka, ed.), Academic Press, New York, 387-394 ]. In this antiviral assay for interferon about 1 unit/mL of interferon is the quantity necessary to produce 50% cytopathic effect. The units are determined with respect to the international reference standard for human interferon-alphaA (Hu-IFN-α? provided by the National Institutes of Health [see Pestka, S. (1986)'Interferon Standards and General Abbreviations,' in Methods in Enzymology Vol. 119 (S. Pestka, ed.), Academic Press, New York, 14-23 ]. The activity was determined with bovine MDBK cells challenged with vesicular stomatitis virus (VSV).
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Content And Storage -80°C
Activity Specific activity = 4.0x10e8 units/mg; Assay used to measure bioactivity: interferon was titrated with the use of the cytopathic effect inhibition assay as described [Rubinstein, S., Familletti, P.C., and Pestka, S. (1981 Convenient Assay for Interferons, J. Virol. 37, 755-758; Familletti, P.C., Rubinstein, S., and Pestka, S. (1981) A Convenient and Rapid Cytopathic Effect Inhibition Assay for Interferon, in Methods in Enzymology, Vol. 78 (S. Pestka, ed.), Academic Press, New York, 387-394]. In this antiviral assay for interferon about 1 unit/mL of interferon is the quantity necessary to produce 50% cytopathic effect. The units are determined with respect to the international reference standard for human interferon-alphaA (Hu-IFN-α? provided by the National Institutes of Health [see Pestka, S. (1986) Interferon Standards and General Abbreviations, in Methods in Enzymology Vol. 119 (S. Pestka, ed.), Academic Press, New York, 14-23]. The activity was determined with bovine MDBK cells challenged with vesicular stomatitis virus (VSV).
Shipping Condition Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Liquid
Protein Subtype Interferons
Research Category Immunology, Virology, Oncology, Signal Transduction
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.