missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-19 Recombinant Protein

Product Code. 10110683
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10110683 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10110683 Supplier Gibco™ Supplier No. PHG0244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 12 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.

FGF19, Fibroblast growth factor 19, is a protein that in humans is encoded by the FGF19 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. The FGF19 gene is mapped to 11q13.3. The deduced 216-amino acid FGF19 protein contains asignal sequence and 2 cysteine residues that are conserved in the FGF family. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. FGF19 stimulates hepatic protein and glycogen synthesis but does not induce lipogenesis. The effects of FGF19 are independent of the activity of either insulin or the protein kinase Akt and, instead, are mediated through a mitogen-activated protein kinase signaling pathway that activates components of the protein translation machinery and stimulates glycogen synthase activity. The orthologous protein in mouse is FGF15, which shares about 50% amino acid identity and has similar functions. Together they are often referred to as FGF15/19.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95750
For Use With (Application) Bioactivity
Formulation 5mM tris with 75mM NaCl and no preservative; pH 7.6
Gene ID (Entrez) 9965
Molecular Weight (g/mol) 21.8 kDa
Name Human FGF-19
pH Range 7.6
Purification Method Purified
Quantity 25 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias FGF; Fgf15; FGF19; FGF-19; Fibroblast growth factor; fibroblast growth factor 15; fibroblast growth factor 19; UNQ334/PRO533
Common Name FGF19
Gene Symbol FGF19
Biological Activity ED50 = 100 - 150 ng/mL; determined by the dose-dependent proliferation of Balb/c 3T3 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human FGF-19 recombinant protein contains 195 amino acids
Content And Storage -20°C
Activity ED50 = 100 - 150 ng/mL; determined by the dose-dependent proliferation of Balb/c 3T3 cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype FGFs (Fibroblast Growth Factors)
Research Category Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis
Product Line Gibco
Purity or Quality Grade ≥95% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.