Learn More
Gibco™ Human CD40 Ligand (TNFSF5) Recombinant Protein
Description
Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute sCD40L in 5-10 mM sodium phosphate, pH 7.5, to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage, preferably desiccated. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P29965 |
| For Use With (Application) | Bioactivity |
| Formulation | 10mM sodium phosphate with no preservative |
| Gene ID (Entrez) | 959 |
| Molecular Weight (g/mol) | 16.3 kDa |
| Name | Human CD40 Ligand (TNFSF5) |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.