missing translation for 'onlineSavingsMsg'
Learn More

BOLA HT Laboratory Screw Joints

Product Code. 10125551
Change view
Click to view available options
Thread Size:
18 in.
25 in.
32 in.
45 in.
Diameter (Metric):
1.6 mm
2 mm
3 mm
9.52 mm
18 mm
20 mm
22 mm
25.4 mm
30 mm
32 mm
Includes:
Black
White
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Diameter (Metric) Includes unitSize
10125551 25 in. 9.52 mm Black Each
12730726 18 in. 1.6 mm White Each
10349331 25 in. 2 mm Black Each
10469001 25 in. 3 mm White Each
10534402 32 in. 9.52 mm Black Each
10561063 32 in. 9.52 mm White Each
10164681 45 in. 18 mm White Each
10669791 45 in. 20 mm White Each
10585932 45 in. 22 mm Black Each
10194821 45 in. 25.4 mm White Each
10736552 45 in. 30 mm White Each
10746552 45 in. 32 mm White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10125551 Supplier BOLA Supplier No. D63068
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HT Laboratory Screw Joints Glass-fibre reinforced PPS cap inner parts made of PPS and PTFE max. temperature 250°C

Black screw cap made of PPS, inner parts consisting of a v-ring (PPS), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (FKM) for tubing diameters under 3 mm (not exposed to the medium). Good chemical resistance, the medium is only exposed to PTFE.

Specifications

Certifications/Compliance FDA
Diameter (Metric) 9.52 mm
Autoclavable Autoclavable
Thread Size 25 in.
Includes Black
Flexibility Rigid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.