Learn More
Honeywell KCL™ Honeywell KCL™ Vitoject™ 890 Gloves
Product Code. 10115374
Change view
Click to view available options
Size:
10
11
Unit Size:
1 pair
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10115374 11 1 pair
10105374 10 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10115374 Supplier Honeywell KCL™ Supplier No. 089011111X
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Made of Viton. Honeywell KCL™ Honeywell KCL™ Vitoject™ 890 Gloves have superb quality and are chemically resistant. The gloves are excellent for working with aggressive chemicals.
  • AQL < 0.65
  • Loose fit allows heat-protecting gloves to be worn underneath
  • Can be re-used after contamination with certain chemicals, after special cleaning process (please consult KCL Laboratories)
  • Can be combined with chemical protective clothing
  • Excellent low degradation
  • Good resistance against polycyclic aromatic hydrocarbons (petrochemicals
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance IFA 1301115
Color Black
Latex Free Yes
Material Viton
Powdered Powder Free
Product Type Gloves
Size 11
Thickness (Metric) 0.7 cm
Length (Metric) 340-360 cm