missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Product Code. 10192982
Change view
Click to view available options
Type:
Adjustable Comb Back
Comb
Preparative Comb
Standard Comb
No. of Wells:
1 Prep, 1 Reference
1 Prep, 2 Reference
10
12
15
20
28
N/A
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
N/A
Volume (Metric) Well:
11.5μL/ mm
112μL/ mm
12.4μL/ mm
120μL/ mm
171μL/ mm
183μL/ mm
2.1μL/ mm
2.7μL/ mm
3.1μL/ mm
4.1μL/ mm
4.3μL/ mm
5.7μL/ mm
5.8μL/ mm
6.2μL/ mm
7.7μL/ mm
8.3μL/ mm
8.6μL/ mm
85μL/ mm
90μL/ mm
N/A
Unit Size:
Each
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type No. of Wells Thickness (Metric) Volume (Metric) Well unitSize
10192982 Standard Comb 15 0.15 cm 8.6μL/ mm Each
11579713 Comb 12 0.1 cm 7.7μL/ mm Each
12084867 Comb 1 Prep, 2 Reference 0.15 cm 171μL/ mm Each
12014707 Comb 1 Prep, 2 Reference 0.07 cm 85μL/ mm Each
12074807 Preparative Comb 1 Prep, 2 Reference 0.1 cm 112μL/ mm Each
12004877 Standard Comb 20 0.07 cm 3.1μL/ mm Each
10660036 Standard Comb 20 0.15 cm 6.2μL/ mm Each
12069807 Standard Comb 20 0.15 cm 6.2μL/ mm Each
12054647 Standard Comb 15 0.07 cm 4.3μL/ mm Each
12074717 Standard Comb 10 0.07 cm 6.2μL/ mm Each
11955348 Standard Comb 12 0.15 cm 11.5μL/ mm Each
11945348 Standard Comb 12 0.07 cm 5.8μL/ mm Each
12064757 Standard Comb 10 0.15 cm 12.4μL/ mm Each
10341135 Standard Comb 10 0.15 cm 12.4μL/ mm Each
10783847 Adjustable Comb Back N/A N/A N/A Each
12089857 Adjustable Comb Back N/A N/A N/A Each
12069817 Comb 1 Prep, 1 Reference 0.07 cm 90μL/ mm Each
12014747 Comb 1 Prep, 1 Reference 0.15 cm 183μL/ mm Each
11569723 Comb 28 0.1 cm 2.7μL/ mm Each
12014807 Comb 1 Prep, 1 Reference 0.1 cm 120μL/ mm Each
12074757 Comb 28 0.07 cm 2.1μL/ mm Each
12079807 Comb 28 0.15 cm 4.1μL/ mm Each
11539723 Comb 20 0.1 cm 4.1μL/ mm Each
12054867 Comb 15 0.1 cm 5.7μL/ mm Each
10142892 Comb 10 0.1 cm 8.3μL/ mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
This item is not returnable. View return policy
Product Code. 10192982 Supplier Hoefer™ Supplier No. SE511151.5
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25 mm, except for the 28-well preparative combs which produce well depths of 15 mm
  • Adjustable comb back (available separately, below) converts 25 mm deep combs to 10 or 15 mm depth

Specifications

For Use With (Equipment) SE 600, SE 400 Series Vertical Electrophoresis System
Type Standard Comb
Material PTFE
No. of Wells 15
Thickness (English) 0.05 in.
Thickness (Metric) 0.15 cm
Volume (Metric) Well 8.6μL/ mm
Width (Metric) 0.57 cm
Depth (English) 0.98 in.
Depth (Metric) 2.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.