missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 10161070
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10161070 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161070 Supplier Invitrogen™ Supplier No. PA515441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HK3 Polyclonal Antibody for Western Blot, IHC (P)

Hexokinases phosphorylate glucose to produce glucose 6 phosphate, thus committing glucose to the glycolytic pathway. In vertebrates there are four major glucose phosphorylating isoenzymes, designated hexokinase I, II, III and IV (glucokinase). Similar to hexokinases I and II, the allosteric enzyme hexokinase III is inhibited by its product glucose 6 phosphate. The catalytic activity of hexokinase III is associated with the C terminus while regulatory function is associated with the N terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HK3
Gene Accession No. P52790, Q3TRM8
Gene Alias ATP:D-hexose 6-phosphotransferase; hexokinase 3; hexokinase 3 (white cell); hexokinase iii; hexokinase type III; Hexokinase3; Hexokinase-3; Hexokinase-C; HK III; HK3; HKIII; HK-III; HXK3; RNU73859
Gene Symbols HK3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 760-789 amino acids from the C-terminal region of human HK3 (Hexokinase III).
Purification Method Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212032, 3101
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.