missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human HIPK2, GST Tag Recombinant Protein

Product Code. 10175653
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
1mg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10175653 10 μg Each
15458703 100 μg 100µg
15458783 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10175653 Supplier Invitrogen™ Supplier No. PV5275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic

HIPK2 is a nuclear serine/threonine kinase that is linked to transcriptional regulation and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9H2X6
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with no preservative
Gene ID (Entrez) 28996
Molecular Weight (g/mol) 73 kDa
Name Human HIPK2, GST Tag
Quantity 10 μg
Gene Alias 1110014O20Rik; B230339E18Rik; cb1009; hHIPk2; HIPK2; homeodomain interacting protein kinase 2; homeodomain-interacting protein kinase 2; Nbak1; Nuclear body-associated kinase 1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; wu:fk56d12
Common Name HIPK2
KinaseFamily DYRK Kinase Family
Protein Length 165-564
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Form Liquid
Protein Subtype Serine/Threonine Kinases
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.