missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HIOC, >98%
GREENER_CHOICE

Product Code. 30001041
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001041 50 mg 50mg
30000834 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001041 Supplier Tocris Bioscience™ Supplier No. 5961/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TrkB receptor agonist; neuroprotectant

Selective TrkB receptor agonist. Protects neurons from excitotoxicity. Protects retinas from light-induced retinal degeneration (LIRD) in vivo. Blood brain barrier and blood-retinal barrier penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314054-36-9
Molecular Formula C16H19N3O3

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 301.34
Purity >98%
Molecular Weight (g/mol) 301.346
Chemical Name or Material N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-2-oxo-3-piperidinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.