missing translation for 'onlineSavingsMsg'
Learn More

Hexanedioic-d8 Acid Dihydrazide, TRC

Product Code. 30322948
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322948 10 mg 10mg
30322844 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322948 Supplier Toronto Research Chemicals Supplier No. TRCA29159610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adipic Acid Dihydrazide-D8
CAS 1202865-37-9
Molecular Formula C6 D8 H6 N4 O2
Quantity 10 mg
Synonym 2,2,3,3,4,4,5,5-Octadeuteriohexanedihydrazide, Hexanedioic-d8 Acid Dihydrazide, Hexanedioic Acid Dihydrazide-D8, Adipic-d8 acid dihydrazide, Adipic Acid Dihydrazide-D8
InChI Formula InChI=1S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)/i1D2,2D2,3D2,4D2
SMILES [2H]C([2H])(C(=O)NN)C([2H])([2H])C([2H])([2H])C([2H])([2H])C(=O)NN
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriohexanedihydrazide
Molecular Weight (g/mol) 182.2503
Formula Weight 182.1619
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.