missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanedioic-d8 Acid Dihydrazide, CDN
Specifications
Specifications
| Chemical Name or Material | Adipic Acid Dihydrazide-D8 |
| CAS | 1202865-37-9 |
| Molecular Formula | C6 D8 H6 N4 O2 |
| Quantity | 0.05 g |
| Synonym | 2,2,3,3,4,4,5,5-Octadeuteriohexanedihydrazide, Hexanedioic-d8 Acid Dihydrazide, Hexanedioic Acid Dihydrazide-D8, Adipic-d8 acid dihydrazide, Adipic Acid Dihydrazide-D8 |
| InChI Formula | InChI=1 S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(C(=O)NN)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)NN |
| IUPAC Name | 2,2,3,3,4,4,5,5-octadeuteriohexanedihydrazide |
| Molecular Weight (g/mol) | 182.2503 |
| Formula Weight | 182.1619 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?