missing translation for 'onlineSavingsMsg'
Learn More

SCAT Round Canisters

Product Code. 10190560 Shop All SCAT Europe Products
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
5 L
10 L
25 L
50 L
60 L
Closure Size:
53B
83B
S65
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10190560 4 L 83B Each
10801511 2 L 53B Each
10830201 5 L S65 Each
10323490 10 L 83B Each
10398319 10 L S65 Each
10650241 25 L S65 Each
17161511 50 L 83B Each
10650431 60 L S65 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10190560 Supplier SCAT Supplier No. 107712
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps or the SCAT Universal Waste Hub 'Jan'.

  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
  • Available in the common threads S65, S70/71 and B83
  • Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents

Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances.

SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.

Specifications

Autoclavable Not Autoclavable
Capacity (English) 1 gal.
Capacity (Metric) 4 L
Closure Size 83B
Color White (Translucent)
Description Round canister 4 L, B83, PE-HD, dimensions (W x H x D): 155 x 338 x 155 mm
Dimensions (L x W x H) 155 x 155 x 338 mm
Disposable Reusable
Material HDPE
Mouth Style B83 Thread
No. per Pack 1 Ea.
Product Type Canister
Shape Round
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.