Learn More
Harmaline, 98+%, Thermo Scientific™
£44.60 - £152.00
Chemical Identifiers
| CAS | 304-21-2 |
|---|---|
| Molecular Formula | C13H14N2O |
| Molecular Weight (g/mol) | 214.268 |
| MDL Number | MFCD00004955 |
| InChI Key | QJOZJXNKVMFAET-UHFFFAOYSA-N |
| Synonym | harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline |
| PubChem CID | 5280951 |
| IUPAC Name | 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole |
| SMILES | CC1=C2C(=C3C=CC(=CC3=N2)OC)CCN1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15404249
|
Thermo Scientific Alfa Aesar
J61699.03 |
1g |
£44.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15414249
|
Thermo Scientific Alfa Aesar
J61699.06 |
£152.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- CNS stimulant; may act through NMDA receptors
- Reversible inhibitor of MAO-A
Chemical Identifiers
| C13H14N2O | |
| MFCD00004955 | |
| harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline | |
| 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole |
Specifications
| 304-21-2 | |
| MFCD00004955 | |
| 14,4613 | |
| QJOZJXNKVMFAET-UHFFFAOYSA-N | |
| 7-methoxy-1-methyl-3,4-dihydro-2H-pyrido[3,4-b]indole | |
| 5280951 | |
| ≥98% |
| C13H14N2O | |
| 207310 | |
| harmaline, dihydroharmine, harmidine, armalin, harmalol methyl ether, o-methylharmalol, 3,4-dihydroharmine, harmine, dihydro, harmalin, 1-methyl-7-methoxy-3,4-dihydro-beta-carboline | |
| CC1=C2C(=C3C=CC(=CC3=N2)OC)CCN1 | |
| 214.268 | |
| 214.27 | |
| Harmaline |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 206-152-6
missing translation for 'rtecsNumber' : UU9800000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.