missing translation for 'onlineSavingsMsg'
Learn More

H3K4me3 Polyclonal Antibody, Invitrogen™

Product Code. 10083864
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083864 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10083864 Supplier Invitrogen Supplier No. 491005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Rabbit Polyclonal Antibody

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post tranlationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tri-Methyl-Histone H3 (Lys4)
Applications ELISA, Immunocytochemistry, Immunofluorescence, Peptide Array, Western Blot
Classification Polyclonal
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% Proclin 300, 0.05% sodium azide
Gene HIST1H3A
Gene Accession No. P68431, P68433
Gene Alias H3K4me3, Histone H3, Histone 1, H3a, Histone H3.1, histone H3/a, HIST1H3A
Gene Symbols H3c1, HIST1H3A
Host Species Rabbit
Immunogen Raised against the region of the histone H3 containing the trimethylated lysine 4 (or [K4me3]), using a KLH-conjugated synthetic peptide.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360198, 8350
Target Species Human, Multi-species, Mouse, Plant, Zebrafish
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.