missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H-2Dk Monoclonal Antibody (15-5-5), FITC

Description
H-2Dk Monoclonal Antibody for Flow
MHC Class I (H-2Dk) antibody reacts with the H-2Dk class I alloantigen. It cross-reacts with H-2Kd and with cells from mice with the H-2f haplotype. The antibody does not react with other (e.g., a, b, p, q, r, s) haplotypes. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
Specifications
| Antigen | H-2Dk |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 15-5-5 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | H2-D1 |
| Gene Accession No. | P01895, P01896, P01897, P01899, P01900, P14426, P14427 |
| Gene Alias | H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H2K1; H2KD; histocompatibility 2, D region locus 1; MHC Class I; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?