missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 583340 dihydrochloride
GREENER_CHOICE

Product Code. 18782544
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782544 10 mg 10mg
18752524 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782544 Supplier Tocris Bioscience™ Supplier No. 2239/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent dual EGFR/ErbB2 inhibitor; orally active

Potent dual EGFR/ErbB2 tyrosine kinase inhibitor (IC50 values are 0.01 and 0.014 μM respectively). Selectively inhibits growth of human tumor cells overexpressing EGFR and ErbB2 (IC50 values are 0.11 μM for inhibition of HN5, N87 and BT474 tumor cell lines vs. > 30 μM for inhibition of non-tumor cell line HFF). Inhibits tumor growth in vivo by ∽ 80% in a murine xenograft model following oral administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173023-85-2
Molecular Formula C28H27Cl3FN5O3S2

Specifications

Quantity 10 mg
Formula Weight 671.03
Purity >97%
Molecular Weight (g/mol) 671.024
Chemical Name or Material GW 583340 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.