missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Guangxitoxin 1E
GREENER_CHOICE

Product Code. 18739248
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18739248 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18739248 Supplier Tocris Bioscience™ Supplier No. 5676/100U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Kv2.1 and Kv2.2 channel blocker

Guangxitoxin 1E is a Kv2.1 and Kv2.2 channel blocker (IC50 values are 1-3 nM). Enhances glucose-stimulated insulin secretion from human islets in vitro, but not from islet cells lacking the Kv2.1 channel. Has no significant effect on plasma insulin, glucagon or blood glucose levels in mice, but increases plasma somatostatin levels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1233152-82-3
Content And Storage Store at -20°C
Description Voltage-gated Potassium Channel Blockers
Molecular Weight (g/mol) Observed M.W.: 3948.61
Quantity 100 μg
Structural Form C178H248N44O45S7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.