missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSK3 alpha/beta Monoclonal Antibody (21A)

Product Code. 10037812
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10037812 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10037812 Supplier Invitrogen™ Supplier No. 44610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

44-610 is an antibody that's produced in mouse against a recombinant Xenopus laevis protein. Has been successfully used ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, and Western Blot procedures.

Glycogen synthase kinase-3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. Since then, it has been shown to be involved in the regulation of a diverse array of cellular functions, including protein synthesis, cell proliferation, cell differentiation, microtubule assembly/disassembly, and apoptosis. GSK3's substrate specificity is unique in that phosphorylation of the substrate only occurs if a phosphoserine or phosphotyrosine is present - four residues C-terminal to the site of GSK phosphorylation. There exists two isoforms of GSK3, GSK3 alpha and GSK3 beta, and they are strictly regulated via phosphorylation. Phosphorylation of GSK3 beta on Ser9 (Ser21 in GSK3alpha) by protein kinase B (PKB) causes its inactivation and is the primary mechanism responsible for growth factor inhibition of this kinase. Activation of GSK3 beta is dependent upon the phosphorylation of Tyr216 (Tyr279 in GSK3 alpha). Upon activation, it has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3 beta also has been shown to phosphorylate aberrant sites on the microtubule associated protein tau, which is critical for the progression of Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3 alpha/beta
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 21A
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene GSK3B
Gene Accession No. P18265, P18266, P49840, P49841, Q2NL51, Q9WV60
Gene Alias 2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B
Gene Symbols GSK3A, GSK3B
Host Species Mouse
Immunogen The antibody was produced in mouse against a recombinant Xenopus laevis protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2931, 2932, 50686, 56637, 606496, 84027
Target Species Human, Mouse, Rat, Frog
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.