missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 2795039
GREENER_CHOICE

Product Code. 18767456
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18767456

Brand: Tocris Bioscience™ 6848/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NADPH oxidase 2 (NOX2) inhibitor

GSK 2795039 is a NADPH oxidase 2 (NOX2) inhibitor (pIC50 value is 5.5 to 6.5, depending on assay performed), which displays selectivity for NOX2 other NOX isoforms, xanthine oxidase and eNOS. GSK 2795039 inhibits reactive oxygen species (ROS) in cellular assays, and displays efficacy in rodent models of paw inflammation and acute pancreatitis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-(2,3-Dihydro-1-methyl-1 H-indol-6-yl)-1-(1-methylethyl)-1 H-pyrrolo[2,3-b]pyridin-4-yl]-1-methyl-1 H-pyrazole-3-sulfonamide
CAS 1415925-18-6
Quantity 25 mg
Target NADPH Oxidase Inhibitors
Molecular Formula C23H26N6O2S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.