Learn More
Karl Hecht™ Ground Glass Joint Cone with Standard Ground
Description
- With extension at the bottom
- DIN 12242
- Manufactured from DURAN™ borosilicate glass 3.3
- Solid rim, form G
- Joint size 14/23
Specifications
Specifications
| Item Description | Form G |
| Type | Ground Glass Joint |
| Joint Size | 14/23 joint |
| Standard Taper Joint No | 14/23 |