missing translation for 'onlineSavingsMsg'
Learn More

GBR Technology™ Grease, 50G, High Vacuum

Product Code. 10162050
Encompass
Change view
Click to view available options
Quantity:
10 × 50g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10162050 10 × 50g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10162050 Supplier GBR Technology™ Supplier No. 970V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High vacuum grease is a silicone-based compound used as a sealant and lubricant for vacuum pressure systems.

  • CAS: 63148-62-9, 70131-67-8, 7631-86-9 E
  • Density: 1.1g/c,3
  • Flash point: >100°C
  • Insoluble in water
  • Store at room temperature in the original container

Specifications

Product Type High Vacuum Grease
Color White to greyish
Packaging Tube
Quantity 10 × 50g
Volume (English) 1.76 oz.
Volume (Metric) 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.