Learn More
Eppendorf™ Grease
Grease pistons when pipette has become extremely dirty. Eppendorf™ Grease is available in 45g tubes, for use with a variety of pipettes.
Brand:  Eppendorf™ 0013019231
96.90 GBP valid until 2026-03-31
BEST PRICE on Promo! Use promo code "28013" to get your promotional price.
Product Code. 10124621
  • ONLINE EXCLUSIVE PRICE
    £102.00
    £94.44 / 45g
    Save £7.56
    7% Off
View all promotions
Description
  • Silicone grease for pipettes
  • Clean pipette with organic solvents; apply grease following cleaning
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Each
45 g