missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Graduation for Wide-Neck Jerry Cans

Product Code. 10157430 Shop All Bürkle Products
Change view
Click to view available options
:
5L wide neck jerry cans
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10157430 5L wide neck jerry cans
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157430 Supplier Buerkle™ Supplier No. 04319051
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For Use With: 5L Jerrican

  • For wide-necked jerrycans
  • Self-adhesive strips with blue graduation marks
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) 5 L Jerrican
Graduations Liters
Product Type Jerry Can Graduation Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.