Learn More
Kartell SpA Graduated Wash Bottle
Product Code. 10062922
Change view
Click to view available options
Capacity (Metric):
250 mL
Color:
Red
Yellow
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
10062922 250 mL Yellow Pack of 10
10676133 250 mL Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10062922 Supplier Kartell SpA Supplier No. 0163706
Only null left
Add to basket
Add to basket
Wide mouth wash bottles, graduated
Wide mouth for easy, safe filling. Color coded caps for easy identification. Graduated. Ingenious cap designed to avoid involuntary dripping due to pressure build up in bottle - where toxic or radioactive material is being used
Specifications
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Yellow
Material Polyethylene
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated Yes
Mouth Wide, Threaded
Shape Round
Fit for Purpose Certified Quality
Product Type Wide Mouth Wash Bottle
Chemical Label Blank
Autoclavable Not Autoclavable