Learn More
Karl Hecht™ Erlenmeyer Flask, Standard Ground
Description
- Duran™ borosilicate glass.
- Graduated.
- Standard ground.
- Reinforced ground neck marked with standard ground size.
- Without stopper.
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass (3.3) |
| Shape | Erlenmeyer |
| Standard Taper Joint | NS 14/23 |
| Graduated | Yes |
| Closure Size | NS 14/23 |