missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight™ 633

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen™ 35512

146.25 GBP valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.


Product Code. 10006103

  • £195.00 / 1mL
Seek Out Great Savings With the Lab of Curiosity
Use promo code 24110
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Product # 35512 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35512 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 633 Amax= 638 nm; Emax= 658 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG (H+L)
Polyclonal
DyLight 633
Goat
Antigen affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1% BSA and 0.02% sodium azide; pH 7.2
Purified Mouse IgG, whole molecule.
1 mL
Secondary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.