Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, Alexa Fluor™ 488-10 nm colloidal gold
Description
This 10 nm colloidal gold conjugate may be used as a probe in immunoblotting, light microscopy, fluorescence microscopy or electron microscopy. The fluorescence of this conjugate can be easily detected by standard techniques, but the ease of visualization of colloidal gold can be greatly improved using silver-enhancement methods, such as those we provide in the LI Silver Enhancement Kit (Cat. no. L-24919).
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | Immunomicroscopy, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 30 μg/mL |
| Conjugate | Alexa Fluor 488-10 nm colloidal gold |
| Formulation | PBS with 5mM sodium azide; pH 7.5 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobin. |
| Purification Method | Purified |
| Quantity | 15 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.