Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Secondary Antibody, AP
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot and IP applications. Antibody Specificity: This antibody reacts with the heavy chains on mouse IgG but not with the light chains common to most mouse immunoglobulins. No antibody was detected against mouse IgM or against non-immunoglobulin serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 4°C until opened. Restore with 1.0 mL distilled water. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG Fc |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Alkaline Phosphatase (AP) |
| Formulation | TBS with 15mg/mL BSA and 0.05% sodium azide; pH 8 |
| Host Species | Goat |
| Purification Method | Affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.