Learn More
Invitrogen™ Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, PE
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31861 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31861 reacts with the heavy chains of mouse IgG subclasses 1, 2a, 2b, and 3, but not with light chains common to most mouse immunoglobulins, but does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 82.0 at 566 nm for a 1% solution for only those R-PE molecules to which at least one molecule of active antibody is bound) Phycoerythrin Source: Seaweed Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. RPE Amax= 490 nm, 545 nm, and 566 nm; Emax= 580 nm. Reconstitute with 1.0 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA.
Specifications
Specifications
| Antigen | Mouse IgG Fc Cross-Adsorbed |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Goat |
| Purification Method | Antigen affinity chromatography |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Afficher plus |
Sécurité et traitement
- Goat anti-Mouse IgG Fc Cross Adsorbed Secondary Antibody, RPEconjugate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: SODIUM AZIDE
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.