missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse Ig (Multiple Adsorption) Secondary Antibody, BD (DUPLICATE)

Product Code. 10026975
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10026975 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10026975 Supplier BD Biosciences Supplier No. 550337

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

X

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat anti-Mouse Ig (Multiple Adsorption)
Classification Polyclonal
Clone POLYCLONAL
Conjugate Biotin
Host Species Goat
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.