missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG (H+L) Secondary Antibody, AP

Product Code. 10016623
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10016623 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10016623 Supplier Invitrogen™ Supplier No. 31310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31310 has been successfully used in Western blot applications. Product # 31310 reacts with the heavy chains of human IgG and with the light chains common to most human immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.0 mL of distilled water (0.6 mg/mL after restoration). Country of Origin: USA.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L)
Applications Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation 0.01M tris HCl with 0.25M NaCl, 15 mg/mL BSA and 0.05% sodium azide; pH 8.0
Host Species Goat
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.