Learn More
Karl Hecht™ Glass Powder Funnel
Product Code. 10165201
Change view
Click to view available options
Diameter (Metric)Top:
5cm
8cm
12cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
10165201 8cm Each
10125251 12cm Each
10258101 5cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10165201 Supplier Karl Hecht™ Supplier No. 44001036
Only null left
Add to basket
Add to basket
Transfer powdered materials easily with this short, wide-stem funnel. Karl Hecht™ Glass Powder Funnel is constructed of AR™ soda glass.
  • Short, wide stem.
  • AR™ soda glass construction.
Specifications
Diameter (Metric)Top 8cm
Material AR-GLAS
For Use With (Application) Powdered materials
Product Type Funnel