Learn More
BOLA GL Bellows
Product Code. 10043381
Change view
Click to view available options
Overall Length (Metric):
104 mm
115 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Overall Length (Metric) unitSize
10043381 115 mm Each
10514212 104 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10043381 Supplier BOLA Supplier No. H90215
Only null left
Add to basket
Add to basket
GL Bellows PTFE. Screw caps made of PPS (black), very flexible (120°)
Two screw caps made of PPS with GL thread and a PTFE/FKM gasket. Bellow with sharp folds made of PTFE. Bellow can be bent up to an angle of 120°, flexible and strainless connection.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Overall Length (Metric) 115 mm
Thread Size 45
Color White