Learn More
Gilson™ Accessory for PIPETMANTM Complete Pipetting System - JimmyTM Microtube Openers
GREENER_CHOICE
Product Code. 10188304 Shop All Gilson Products
Change view
Click to view available options
Unit Size:
Pack of 3
Product Code. unitSize
10188304 Pack of 3
1 options
Product Code. 10188304 Supplier Gilson™ Supplier No. F144983
Only null left
Add to basket
Add to basket
JimmyTM Microtube Openers for use with PIPETMAN ClassicTM and PIPETMANTM Neo pipets.
  • For opening both snap-cap and screw-cap microtubes
  • Increases pipeting efficiency, lessens fatigue associated with manual tube opening and eliminates common mode of contamination
TRUSTED_SUSTAINABILITY
Specifications
For Use With All Pipettes with Tips Mounted
Material Polypropylene
Type Microtube Opener
For Use With (Equipment) PIPETMAN™ Complete Pipetting System
Quantity 3 Pk.