missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL13 (BLC/BCA-1) Recombinant Protein
Description
Human CXCL13 (BLC/BCA-1) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Recombinant human BCA-1 is intended for use in cell culture applications. BCA-1 is involved in chemotaxis of B-lymphocytes.
- This product is supplied lyophilized, carrier-free
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Specifications
| Accession Number | AF030636, AK156025 |
| For Use With (Application) | Cell Culture |
| Gene ID (Entrez) | AF030636, AK156025 |
| Molecular Weight (g/mol) | 10.1 kDa |
| Purification Method | Sequential Chromatography |
| Gene Alias | BCA-1 (CXCL13), GRO beta, GROB, Gro b, CXCL13 |
| Cell Type | B Cells |
| Gene Symbol | CXCL13 |
| Protein Length | 87 aa |
| Species | Human |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?