missing translation for 'onlineSavingsMsg'
Learn More

GGTI298 Trifluoroacetate, MedChemExpress

Product Code. 16491559
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491559 1 mg 1mg
16489417 5 mg 5mg
16443506 25 mg 25mg
16412947 50 mg 50mg
16406888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491559 Supplier Medchem Express Supplier No. HY15871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM; little effect on Ha-Ras with IC50 of >20 μM.

GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM

Specifications

Chemical Name or Material GGTI298 Trifluoroacetate
CAS 1217457-86-7
Color White
Purity Grade Notes Research
Molecular Formula C29H34F3N3O5S
Quantity 1 mg
Solubility Information DMSO : 150 mg/mL (252.67 mM; Need ultrasonic and warming)
SMILES CC(C)C[C@@H](C(OC)=O)NC(C1=CC=C(NC[C@@H](N)CS)C=C1C2=C3C=CC=CC3=CC=C2)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 593.66
Formula Weight 593.66
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.