missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Genie-Vortex 2 Replacement Eccentric with Clamp Assembly

Product Code. 11645371
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie 2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11645371 Vortex-Genie 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11645371 Supplier Scientific Industries Supplier No. 0K0236904
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Genie-Vortex 2 Eccentric with Clamp Assembly is a replacement kit to be used with a Genie-Vortex 2 Motor Assembly.

  • This kit contains the Eccentric with Clamp Assembly, which includes the Eccentric, Bearing Retainer, and Eccentric Clamp
  • This assembly is used on the Genie-Vortex 2 Motor Assembly

Specifications

Product Type Eccentric Replacement
Color Black
For Use With (Application) Motor Assembly
For Use With (Equipment) Vortex-Genie 2
Includes Eccentric, Bearing Retainer, and Eccentric Clamp
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.