missing translation for 'onlineSavingsMsg'
Learn More

Robu Gas-Washing-Bottle, FD 100ml, Por. 2, NS 29/32

Brand:  Robu 40102

Gas-Washing-Bottle, FD 100ml, Por. 2, NS 29/32

Product Code. 10113981

  • ONLINE EXCLUSIVE PRICE
    £74.85
    £73.54 / Each
    Save £1.31
    1% Off
Only null left
Estimated Shipment: 08-04-2026
Add to basket
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.