missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Gas-permeable Foil

Product Code. 10114571
Change view
Click to view available options
Material:
Aluminum
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
10114571 Aluminum Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10114571 Supplier Macherey-Nagel™ Supplier No. 740675
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Foil to cover square‐well blocks during incubation of bacterial cultures.

NucleoSpin™ 96 Blood is designed for the parallel isolation of genomic DNA from whole blood. After lysis of blood cells genomic DNA is selectively bound to a silica membrane while contaminants are washed away effectively. Elution is possible even under vacuum without cross-contamination. Due to its plate design and the possibility to perform the complete procedure under vacuum NucleoSpin™ 96 Blood can be easily adapted to common liquid handling instruments for fully automated processing without manual interactions. Kit components: NucleoSpin™ Blood binding plate, MN wash plate, buffers, proteinase K, 15mL tubes, cap strips, square-well block, rack with tube strips.

Required hardware:

Vacuum processing - the NucleoSpin 96 blood kits can be used manually with the NucleoVac 96 vacuum manifold. For vacuum processing of the 8-well strips the reusable starter set A, containing column holders A and dummy strips is required. For automation on laboratory platforms with standard 96-well vacuum chambers the use of the starter set A is also required.
  • Silica membrane technology
  • Yield: 4μg to 6μg
  • Sample material: ≤200uL whole blood/2 x 106 cultured cells
  • Elution volume: 100μL
  • Binding capacity: 20μg
  • Preparation time: 70 min./plate
  • Format: 96 well plates
  • Fragment size: 300bp to 30kb
  • No use of organic solvents Suitable for whole blood (human or animal); Whole blood treated with citrate, EDTA, heparin.

Specifications

Material Aluminum
No. per Pack 50
For Use With (Application) For The Taping Of The Square Well Plate
Product Type Gas Permeable Foil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.