Learn More
Lenz Laborglasintrumente Gas Inlet Tubes, Straight
Product Code. 10070191
Change view
Click to view available options
Standard Taper Joint:
Cone NS 24/29
Cone NS 29/32
NS 19/26
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
10070191 Cone NS 29/32 Each
11836114 NS 19/26 Each
10284001 Cone NS 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10070191 Supplier Lenz Laborglasintrumente Supplier No. 05080004
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Straight air leak tubes
  • Built-in length: 200 mm
  • With NS cone and thin capillary (easily to draw by burner)
  • Made of DURAN™ tubing
Specifications
Material Duran Glass
Standard Taper Joint Cone NS 29/32