Learn More
Testo™ Frozen food probe
Product Code. 10084481
Change view
Click to view available options
Description:
Frozen food probe
Unit Size:
Each
Product Code. Description unitSize
10084481 Frozen food probe Each
1 options
Product Code. 10084481 Supplier Testo™ Supplier No. 0603 3292
Only null left
Add to basket
Add to basket
Specifications
Description Frozen food probe
Type Thermocouple type T
For Use With (Application) Measuring the core temperature of frozen food
Includes 1.7 m plug-in cable