missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FRK (PTK5), GST Tag Recombinant Protein

Product Code. 15488613
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488613 100 μg 100µg
10056892 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488613 Supplier Invitrogen™ Supplier No. PR7729A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42685
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 2444
Molecular Weight (g/mol) 86.3 kDa
Name Human FRK (PTK5), GST Tag
Quantity 100 μg
Regulatory Status RUO - research use only
Gene Alias B-cell src-homology tyrosine kinase; beta-cell Src-homology tyrosine kinase; BSK; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; FRK; fyn related Src family tyrosine kinase; fyn-related kinase; fyn-related Src family tyrosine kinase; GASK; Gastrointestinal-associated kinase; gastrointestinal-associated tyrosine kinase; GTK; intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; tyrosine-protein kinase FRK
Common Name FRK
KinaseFamily SRC Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.