missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forskolin, MedChemExpress
Description
Forskolin is also an inducer of intracellular cAMP formation. Forskolin induces differentiation of various cell types and activates pregnane X receptor (PXR) and FXR. Forskolin exerts a inotropic effect on the heart, and has platelet antiaggregatory and antihypertensive actions. Forskolin also induces autophagy.
Specifications
Specifications
| Chemical Name or Material | Forskolin |
| CAS | 66575-29-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H34O7 |
| Quantity | 200 mg |
| Synonym | Coleonol Colforsin |
| Solubility Information | DMSO : 100 mg/mL (243.61 mM; Need ultrasonic) |
| SMILES | O=C1[C@@]2(O)[C@]([C@@H](OC(C)=O)[C@@H](O)[C@@]3([H])C(C)(C)CC[C@H](O)[C@@]32C)(C)O[C@@](C)(C=C)C1 |
| Molecular Weight (g/mol) | 410.5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?